missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bis(nonafluorobutanesulfonyl)imide 95.0+%, TCI America™

$130.48 - $393.32

Chemical Identifiers

CAS 119229-99-1
Molecular Formula C8HF18LiNO4S2
Molecular Weight (g/mol) 588.13
MDL Number MFCD06200837
InChI Key SVSJFMZWEBYTRE-UHFFFAOYSA-N
Synonym Bis(nonafluorobutanesulfonyl)imide Lithium Salt
PubChem CID 131868707
IUPAC Name lithium;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)butane-1-sulfonamide
SMILES [Li].C(C(C(F)(F)S(=O)(=O)NS(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L03071G
View Documents
TCI America
L03071G
1 g N/A
Only null left
 
L03075G
View Documents
TCI America
L03075G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

119229-99-1
588.13
SVSJFMZWEBYTRE-UHFFFAOYSA-N
131868707
[Li].C(C(C(F)(F)S(=O)(=O)NS(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F
C8HF18LiNO4S2
MFCD06200837
Bis(nonafluorobutanesulfonyl)imide Lithium Salt
lithium;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)butane-1-sulfonamide

Specifications

119229-99-1
C8HF18LiNO4S2
1 g
Bis(nonafluorobutanesulfonyl)imide Lithium Salt
[Li].C(C(C(F)(F)S(=O)(=O)NS(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F
588.13
587.12
Crystalline Powder
White
MFCD06200837
1759
SVSJFMZWEBYTRE-UHFFFAOYSA-N
lithium;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)butane-1-sulfonamide
131868707
≥95.0% (T)
Lithium Bis(nonafluorobutanesulfonyl)imide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'