missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Catalog No. p-7131909
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02815G 5 g
L028125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02815G Supplier TCI America Supplier No. L02815G

Chemical Identifiers

CAS 171611-11-3
Molecular Formula F2LiNO4S2
Molecular Weight (g/mol) 187.06
MDL Number MFCD28126786
InChI Key VDVLPSWVDYJFRW-UHFFFAOYSA-N
Synonym LiFSI
PubChem CID 86277430
IUPAC Name lithium(1+) bis(fluorosulfonyl)azanide
SMILES [Li+].FS(=O)(=O)[N-]S(F)(=O)=O

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 187.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Bis(fluorosulfonyl)imide
EINECSNumber (1)-1247
TSCA Yes