missing translation for 'onlineSavingsMsg'
Learn More

Lithium benzoate, 99+%

Catalog No. AAA1657130
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1657130 250 g
AAA165710B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1657130 Supplier Thermo Scientific Chemicals Supplier No. A1657130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 553-54-8 | C7H5LiO2 | 128.06 g/mol

In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

Solubility
Solubility in water is increased by sodium benzoate

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 553-54-8
Molecular Formula C7H5LiO2
Molecular Weight (g/mol) 128.06
MDL Number MFCD00035540
InChI Key LDJNSLOKTFFLSL-UHFFFAOYSA-M
Synonym lithium benzoate, benzoic acid, lithium salt, benzoic acid lithium salt, unii-r9z042z19e, benzoic acid, lithium salt 1:1, lithium benzoate nf, lithiumbenzoate, lithotab benzoate, lithium 1+ ion benzoate, ksc491a5n
PubChem CID 2724073
SMILES [Li+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point >300°C
Odor Odorless
Quantity 250 g
Beilstein 3598089
Merck Index 14,5523
Solubility Information Solubility in water is increased by sodium benzoate
IUPAC Name lithium;benzoate
Formula Weight 128.06
Percent Purity ≥99%
Chemical Name or Material Lithium benzoate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 209-042-6
RTECSNumber OJ5720000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only