missing translation for 'onlineSavingsMsg'
Learn More

Lithium arsenate, 99%

Catalog No. AA3935014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3935014 25 g
AA3935006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3935014 Supplier Thermo Scientific Chemicals Supplier No. 03935014
Only null left

CAS: 13478-14-3 | AsLi3O4 | 159.738 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13478-14-3
Molecular Formula AsLi3O4
Molecular Weight (g/mol) 159.738
MDL Number MFCD00053998
InChI Key VKJKOXNPYVUXNC-UHFFFAOYSA-K
Synonym lithium arsenate, trilithium arsenate, trilithium arsorate, arsenic acid h3aso4 , trilithium salt, acmc-20akn0, aso4.3li, arsenic acid h3aso4 , lithium salt 1:3, trilithium 1+ ion arsenate, arsenic acid h3aso4 ,lithium salt 1:3
PubChem CID 166828
IUPAC Name trilithium;trioxido(oxo)-$l^{5}-arsane
SMILES [Li+].[Li+].[Li+].[O-][As](=O)([O-])[O-]

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN1557
Solubility Information Soluble in dilute acetic acid; Insoluble in water
Formula Weight 159.74
Percent Purity 99%
Density 3.07 g/mL
Chemical Name or Material Lithium arsenate
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 236-773-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only