missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum oxide

Catalog No. p-4824792
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1341218 50 g
AA1341230 250 g
AA13412A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1341218 Supplier Thermo Scientific Chemicals Supplier No. 01341218
Only null left

CAS: 12003-67-7 | AlLiO2 | 65.92 g/mol

It is used as an inert electrolyte support material in molten carbonate fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12003-67-7
Molecular Formula AlLiO2
Molecular Weight (g/mol) 65.92
MDL Number MFCD00016172
InChI Key YQNQTEBHHUSESQ-UHFFFAOYSA-N
Synonym lithium aluminate, lithium metaaluminate, aluminum lithium dioxide, aluminate alo21-, lithium, lithium aluminum spinal, lithium aluminate lialo2, hsa 10, aluminum lithium oxide, aluminum lithium oxide lialo2, lithium aluminum oxide lialo2
PubChem CID 123268
IUPAC Name lithium;oxido(oxo)alumane
SMILES [Li+].[O-][Al]=O

Specifications

Melting Point >1625°C
Color White
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 65.92
Odor Odorless
Density 2.62
Chemical Name or Material Lithium aluminum oxide
EINECSNumber 234-434-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only