missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium aluminum oxide
Description
It is used as an inert electrolyte support material in molten carbonate fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >1625°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 1 kg |
| Odor | Odorless |
| Density | 2.62 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 65.92 |
| Chemical Name or Material | Lithium aluminum oxide |
Safety and Handling
| EINECSNumber | 234-434-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only