missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrure de lithium-aluminium (poudre) 95,0+%, TCI America™
Spécifications
Spécifications
| Color | Blanc |
| Melting Point | 125°C |
| Quantity | 25 g |
| Form | Crystalline Powder |
| UN Number | 1410 |
| Formula Weight | 37.95 |
| Percent Purity | ≥98.0% (T) |
| Chemical Name or Material | Lithium Aluminum Hydride (Powder) |
Sécurité et manipulation
| EINECSNumber | (1)-0016 |
| RTECSNumber | BD0100000 |
| TSCA | Yes |