missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Lithium Aluminum Hydride

Catalog No. 1998771KG
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1998771KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1998771KG Supplier Sigma Aldrich Supplier No. 1998771KG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium Aluminum Hydride

Specifications

CAS 16853-85-3
Melting Point 125°C (decomposition)
Molecular Formula H4AlLi
Linear Formula LiAlH4
MDL Number MFCD00011075
Quantity 1 kg
Synonym LAH; Lithium alanate; Lithium tetrahydroaluminate
Molecular Weight (g/mol) 37.95 g/mol