missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Aluminum Di-n-butylamide, 0.16M soln. in 1,2-Dimethoxyethane
Description
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.
Notes
Air, heat and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, air, heat and water.
Lithium aluminum di-n-butylamide is used as a reagent for the determination of hydroxyl groups in silanols.
Notes
Air, heat and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, air, heat and water.
Specifications
Specifications
| Melting Point | -54°C |
| Density | 0.86 |
| Linear Formula | LiAl[N(CH2CH2CH2CH3)2]4 |
| Quantity | 100 mL |
| UN Number | UN1993 |
| Sensitivity | Air and moisture sensitive |
| Formula Weight | 546.87 |
| Chemical Name or Material | Lithium aluminum di-n-butylamide |
Safety and Handling
| Hazard Category | H225-H315-H319-H332-H335-H360FD |
| Hazard Statement | GHS H Statement H226-H315-H319 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only