missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Lithium aluminate
Description
Lithium aluminate
Specifications
Specifications
| CAS | 12003-67-7 |
| Quantity | 250 g |
| Molecular Formula | AlLiO2 |
| Linear Formula | LiAlO2 |
| MDL Number | MFCD00016172 |
| Synonym | Aluminum lithium oxide |
| Molecular Weight (g/mol) | 65.92 |
| Density | 2.615 g/mL (at 25°C) |