missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetylide ethylenediamine complex, tech. 85%, Thermo Scientific Chemicals

Catalog No. aal1240114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1240114 25 g
AAL1240122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL1240114 Supplier Thermo Scientific Chemicals Supplier No. L1240114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Lithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.

Solubility
Soluble in ethylene diamine, n-propyl amine, n-butyl amine, pyridine, dimethyl formamaide, diglyme, dioxan and dimethylsulfoxide.

Notes
Moisture and light sensitive. Store in cool place. Incompatible with alcohols, acids, oxidizing agents, water. It reacts with water to form lithium hydroxide and acetylene, which is a flammable gas. Over time, pressure may increase causing containers to burst. Handle and open container with care.

Chemical Identifiers

CAS 6867-30-7
Molecular Formula C4H9LiN2
Molecular Weight (g/mol) 92.07
MDL Number MFCD00013183
InChI Key QJQWXTYPTBEPGS-UHFFFAOYSA-N
Synonym lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine
PubChem CID 2724010
IUPAC Name lithium;ethane-1,2-diamine;ethyne
SMILES [Li]C#C.NCCN

Specifications

Melting Point ∽76°C (decomposition)
Density 1.04
Odor Amine-like
Linear Formula HC≡CLi·NH2CH2CH2NH2
Quantity 25 g
UN Number UN3131
Beilstein 3593824
Sensitivity Moisture sensitive
Solubility Information Soluble in ethylene diamine,n-propyl amine,n-butyl amine,pyridine,dimethyl formamaide,diglyme,dioxan and dimethylsulfoxide.
Formula Weight 92.07
Percent Purity 90%
Grade Technical
Chemical Name or Material Lithium acetylide ethylenediamine complex
Show More Show Less
Hazard Category H260-H302-H314-H335
Hazard Statement GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
EINECSNumber 229-967-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only