missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetoacetate 97.0+%, TCI America™

$205.05 - $669.19

Chemical Identifiers

CAS 11-2-3483
Molecular Formula C4H5LiO3
Molecular Weight (g/mol) 108.02
MDL Number MFCD00013060
InChI Key UTLRZTUJSMCBHB-UHFFFAOYSA-M
Synonym Acetoacetic Acid Lithium Salt, Lithium 3-Oxobutyrate, 3-Oxobutyric Acid Lithium Salt
PubChem CID 2724246
IUPAC Name lithium;3-oxobutanoate
SMILES [Li+].CC(=O)CC(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A14781G
View Documents
TCI America
A14781G
1 g
Each for $205.05
Only null left
 
A14785G
View Documents
TCI America
A14785G
5 g
Each for $669.19
Only null left
 
Description

Description

Specifications

Chemical Identifiers

11-2-3483
108.02
UTLRZTUJSMCBHB-UHFFFAOYSA-M
2724246
[Li+].CC(=O)CC(=O)[O-]
C4H5LiO3
MFCD00013060
Acetoacetic Acid Lithium Salt, Lithium 3-Oxobutyrate, 3-Oxobutyric Acid Lithium Salt
lithium;3-oxobutanoate

Specifications

White
Crystalline Powder
C4H5LiO3
Acetoacetic Acid Lithium Salt, Lithium 3-Oxobutyrate, 3-Oxobutyric Acid Lithium Salt
[Li+].CC(=O)CC(=O)[O-]
108.02
108.02
Lithium Acetoacetate
11-2-3483
1 g
MFCD00013060
UTLRZTUJSMCBHB-UHFFFAOYSA-M
lithium;3-oxobutanoate
2724246
≥97.0% (T)
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'