missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetoacetate 97.0+%, TCI America™

Catalog No. A14781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14781G 1 g
A14785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14781G Supplier TCI America Supplier No. A14781G
  • Appearance: White to almost white powder to crystal
  • Purity (≥97.0% T): Determined by nonaqueous titration
  • Molecular Formula: C4H5LiO3
  • Molecular Weight: 108.02
  • Solubility in Water: Very faint turbidity
  • Storage Conditions: Store under inert gas in a freezer (<0°C)
  • Sensitivity: Moisture sensitive and heat sensitive

Chemical Identifiers

CAS 11-2-3483
Molecular Formula C4H5LiO3
Molecular Weight (g/mol) 108.02
MDL Number MFCD00013060
InChI Key UTLRZTUJSMCBHB-UHFFFAOYSA-M
Synonym Acetoacetic Acid Lithium Salt, Lithium 3-Oxobutyrate, 3-Oxobutyric Acid Lithium Salt
PubChem CID 2724246
IUPAC Name lithium;3-oxobutanoate
SMILES [Li+].CC(=O)CC(=O)[O-]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 g
Linear Formula CH3COCH2COOLi
Solubility Information Very Faint Turbidity
Formula Weight 108.02
Percent Purity ≥97.0% (T)
Manufacturing Origin Japan
Identification Passes Test
Sensitivity Moisture Sensitive, Heat Sensitive
Appearance White to Almost white powder to crystal
Chemical Name or Material Lithium Acetoacetate
Show More Show Less
TSCA No
Recommended Storage Freezer