missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetate 98.0+%, TCI America™

Catalog No. L0191500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0191500G 500 g
L019125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0191500G Supplier TCI America Supplier No. L0191500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 546-89-4
Molecular Formula C2H3LiO2
Molecular Weight (g/mol) 65.98
MDL Number MFCD00013057
InChI Key XIXADJRWDQXREU-UHFFFAOYSA-M
Synonym lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
PubChem CID 3474584
ChEBI CHEBI:63045
IUPAC Name lithium(1+) acetate
SMILES [Li+].CC([O-])=O

Specifications

Melting Point 283°C
Color White
Quantity 500 g
Formula Weight 65.98
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium Acetate
EINECSNumber (2)-0692
RTECSNumber AI5450000
TSCA Yes