Learn More
Lithium acetate dihydrate, 99%
CAS: 6108-17-4 | C2H7LiO4 | 102.014 g/mol
Supplier: Thermo Scientific Chemicals A179210B
Description
Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
Solubility
Soluble in water and ethanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Lithium acetate dihydrate | |
| 54°C to 57°C | |
| C2H7LiO4 | |
| 1000 g | |
| 14,5521 | |
| Soluble in water and ethanol. | |
| [Li+].CC(=O)[O-].O.O | |
| 102.014 | |
| 102.01 (65.98 Anhydrous) |
| 6108-17-4 | |
| 1.3 | |
| MFCD00066949 | |
| 3564320 | |
| lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate | |
| IAQLJCYTGRMXMA-UHFFFAOYSA-M | |
| lithium;acetate;dihydrate | |
| 23666338 | |
| 99% |
Chemical Identifiers
| 6108-17-4 | |
| 102.014 | |
| IAQLJCYTGRMXMA-UHFFFAOYSA-M | |
| 23666338 | |
| [Li+].CC(=O)[O-].O.O |
| C2H7LiO4 | |
| MFCD00066949 | |
| lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate | |
| lithium;acetate;dihydrate |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 208-914-3
RTECSNumber : AI5450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only