missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetate, Crystal, Spectrum™ Chemical

Catalog No. p-7165784
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611294 125 g Poly Bottle
18611295 500 g Poly Bottle
18611297 2.5 kg Poly Bottle
18611296 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611294 Supplier Spectrum Chemical Mfg Cor Supplier No. L1100125GM
Only null left

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.01
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
IUPAC Name lithium(1+) acetate dihydrate
SMILES [Li+].O.O.CC([O-])=O

Specifications

pH 5.0 to 8.0
CAS Max % 1
Quantity 125 g
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.