missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetate, Crystal, Spectrum™ Chemical

Catalog No. 18611297
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611297 2.5 kg Poly Bottle
18611294 125 g Poly Bottle
18611295 500 g Poly Bottle
18611296 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611297 Supplier Spectrum Chemical Mfg Cor Supplier No. L11002.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity laboratory reagent used as a buffer for gel electrophoresis of RNA and DNA.

  • Grade: Reagent
  • Appearance: Odorless white powder
  • Molecular formula: LiC2H3O2·2H2O
  • Molecular Weight: 102.01
  • CAS Number: 6108-17-4
  • Solubility: Soluble in water
  • Anticorrosion properties
  • pH (5% solution): 5.0 to 8.0
  • Chloride (Cl): 0.002%
  • Sulfate: 0.005%
  • Heavy metals (as Pb): 0.001%
  • Iron (Fe): 0.001%
  • Insolubles: 0.005%
  • Used as a buffer for gel electrophoresis of RNA and DNA

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.01
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
IUPAC Name lithium(1+) acetate dihydrate
SMILES [Li+].O.O.CC([O-])=O

Specifications

pH 5.0 to 8.0
CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 2.5 kg
Linear Formula CH3COOLi⋅2H2O
MDL Number MFCD00149725
Solubility Information Soluble in water
PubChem CID 11966657
Formula Weight 102.01
Grade Reagent
Concentration or Composition (by Analyte or Components) Sulfate: 0.005 %, Insolubles: 0.005 %, Chloride: 0.002 %, Heavy Metals (as Pb): 0.001 %, Iron: 0.001 %
Packaging Poly Bottle
Appearance Crystals
Chemical Name or Material Lithium Acetate, Crystal
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.