missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Acetate, Crystal, Spectrum™ Chemical
Specifications
Specifications
| Color | White |
| pH | 5.0 to 8.0 |
| CAS Max % | 1 |
| Physical Form | Solid |
| Quantity | 125 g |
| Linear Formula | CH3COOLi⋅2H2O |
| MDL Number | MFCD00150395 |
| Solubility Information | Soluble in water |
| PubChem CID | 166826359 |
| Formula Weight | 102.01 |
| Show More |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.