missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate, anhydrous, 99%, extra pure

Catalog No. p-4380064
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC268640010 1 kg
AC268640050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268640010 Supplier Thermo Scientific Chemicals Supplier No. 268640010
Only null left

CAS: 546-89-4 | C2H3LiO2 | 65.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 546-89-4
Molecular Formula C2H3LiO2
Molecular Weight (g/mol) 65.98
MDL Number MFCD00013057
InChI Key XIXADJRWDQXREU-UHFFFAOYSA-M
Synonym lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
PubChem CID 3474584
ChEBI CHEBI:63045
IUPAC Name lithium;acetate
SMILES [Li+].CC([O-])=O

Specifications

Melting Point 280.0°C to 285.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula CH3COOLi
Quantity 1 kg
Beilstein 02,107
Merck Index 15,5577
Solubility Information Solubility in water: 408g/L (20°C). Other solubilities: 667g/L (40°C),938g/L (50°C),freely soluble in alcohol
Formula Weight 65.99
Percent Purity 99%
Grade Extra Pure
Insoluble Matter 0.1% max. (in water)
Physical Form Crystals or Powder
Chemical Name or Material Lithium acetate, anhydrous
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 208-914-3

RUO – Research Use Only