missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lissamine Green B, pure

Catalog No. AC190100250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190100250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC190100250 Supplier Thermo Scientific Chemicals Supplier No. 190100250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3087-16-9 | C27H25N2NaO7S2 | 576.614 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3087-16-9
Molecular Formula C27H25N2NaO7S2
Molecular Weight (g/mol) 576.614
MDL Number MFCD00012117
InChI Key WDPIZEKLJKBSOZ-UHFFFAOYSA-M
Synonym Acid Green 50, C.I. 44090, Wool Green S
PubChem CID 91525
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-3-hydroxynaphthalene-2,7-disulfonate
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+]

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV_Vis 50% min.
Beilstein 14, II, 514
Color Black or Brown
Quantity 25 g
Physical Form Powder
Formula Weight 576.6
Grade Pure
Chemical Name or Material Lissamine Green B
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only