missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ashton Pumpmatic Liquid Dispensing System
Description
- Rapid, drop-free fluid sampling, transferring, and dispensing
- Speeds and simplifies fluid sampling from 1 mL to 10 mL per sample
- Serological pipette/dispenser combination offers accuracy of a reusable at the cost of a disposable
- Controlled rate of sampling and dispensing delivers accurate volumes without drips or spills
- Hazardous liquids do not need to be closer than arm's length
- Transparent, totally closed system
- Features accurate, clearly marked, easily-read graduations
- Smooth-action syringe and plunger with Luer Lok™ permit rapid, one-handed sampling and dispensing
- Lightweight, unbreakable, clear polystyrene
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Polystyrene, Polypropylene |
| Sterility | Sterile |
| Graduated | Yes |
| Packaging Type | Individually Wrapped |
| Color | Clear |
| Volume (Metric) | 1 mL |
| Type | Serological Pipet, Single-use |
| Disposable | Single-use |
| Fit for Purpose | Sterile |