missing translation for 'onlineSavingsMsg'
Learn More

KOVA™ Liqua-Trol™ Liquid Controls

Catalog No. p-109704
Change view
Click to view available options
Type:
Liqua-Trol Liquid Control with HCG
Level:
Leve I (Abnormal) and II (Normal)
Quantity:
6 x 15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Level Quantity
22749755 Liqua-Trol Liquid Control with HCG Leve I (Abnormal) and II (Normal) 6 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22749755 Supplier KOVA™ Supplier No. K87112E
Only null left

Ready-to-use bi-level liquid control for use with all major brands of urine chemistry dipsticks

 

KOVA Liqua-Trol l control helps to validate the performance of urine chemistry test strips and readers and to focus on microscopic sediment analysis. Choose the KOVA controls that best meet the needs of  your lab. For ready-to-use Urinalysis Controls, choose bi-level liquid control for all major brands of urine chemistry dipsticks.


  • Ready-to-use liquid human urine control
  • External quality control of physical, chemical and microscopic examination of urine specimens
  • Available with or without microscopics
  • Three levels of controls  to monitor complete decision range for urine strip chemistries
  • Value assignments available on all major systems for visual and instrument analysis
  • Stability: stable until the expiration date stated on the label, when stored between 2° and 8°C. Do not freeze. Liqua-Trol can be stored at room temperature (20° to 25°C) for up to 30 days. Controls used at room temperature are for reagent strip and hCG testing only. Label the bottle with the date it was originally brought to room temperature to ensure proper performance.

Recommended Storage

Stable until expiration date stated on the label, when stored between 2° to 8°C. Do not freeze. Liqua-Trol can be stored at room temperature (20° to 25°C) for up to 30 days.

Specifications

Certifications/Compliance CLSI and CLIA, ANSI Z400.5-2004
Color Yellow, Green or Amber
Content And Storage 2°C to 8°C
Description Liqua-Trol Liquid Control with HCG
For Use With (Equipment) Urine chemistry dipsticks
Type Liqua-Trol Liquid Control with HCG
Intended Use Validate performance of urine chemistry test strips and readers and to focus on microscopic sediment analysis
Level Leve I (Abnormal) and II (Normal)
Quantity 6 x 15 mL
Form Liquid