missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Lionheart FX Automated Microscope
GREENER_CHOICE

Catalog No. btlfxwsn
Change view
Click to view available options
Camera:
CCD
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Camera
BTLFXWSN CCD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTLFXWSN Supplier Agilent BioTek Supplier No. LFXWSN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. For Research Use Only. Not for use in diagnostic procedures.

Lionheart FX automated microscope is a compact system for a broad range of imaging workflows. It offers up to 60x air; 60x and 100x oil immersion magnification, with fluorescence, brightfield, color brightfield, and phase contrast channels for maximum application reach

An optional environment control cover provides incubation to 40°C and effective containment for CO2/O2 control. A humidity chamber optimizes conditions for long-term live cell imaging applications, and an available dual reagent injector facilitates rapid kinetic assays.

Specifications

Type Cell Imager
Camera CCD
For Use With (Application) Live Cell Imaging
Model LFX
Light Source High Power LED
Software Gen5 Microplate Reader and Imager
Depth (English) 18.3 in.
Depth (Metric) 46.5 cm
Height (English) 14 in.
Height (Metric) 35.6 cm
Width (English) 17.9 in.
Width (Metric) 45.5 cm
Certifications/Compliance CE and TUV marked. RoHS compliant.
Warranty 1 Year
Frequency 50/60 Hz
Voltage 110/220 V
Show More Show Less