missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linoleic Acid 97.0+%, TCI America™
Supplier: TCI America L012425G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C18H32O2 | |
| MFCD00064241 | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| CHEBI:17351 | |
| CCCCCC=CCC=CCCCCCCCC(=O)O |
Specifications
| Linoleic Acid | |
| -5°C | |
| 177°C | |
| 25 g | |
| linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid | |
| CCCCCC=CCC=CCCCCCCCC(=O)O | |
| 280.452 | |
| CHEBI:17351 | |
| ≥97.0% (GC,T) |
| 60-33-3 | |
| Yellow | |
| C18H32O2 | |
| MFCD00064241 | |
| OYHQOLUKZRVURQ-HZJYTTRNSA-N | |
| (9Z,12Z)-octadeca-9,12-dienoic acid | |
| 5280450 | |
| 280.45 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0609
RTECSNumber : RF9990000
TSCA : Yes
Recommended Storage : Refrigerator