missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linoleic acid, 99%

CAS: 60-33-3 | C18H32O2 | 280.45 g/mol

Supplier:  Thermo Scientific Chemicals 215040250

Quantity 25 mL
Packaging Glass bottle

 View more versions of this product

Catalog No. AC215040250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60-33-3
280.45
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
CHEBI:17351
CCCCCC=CCC=CCCCCCCCC(=O)O
C18H32O2
OYHQOLUKZRVURQ-HZJYTTRNSA-N
5280450
(9Z,12Z)-octadeca-9,12-dienoic acid

Specifications

Linoleic acid
-5.0°C
0.9000g/mL
>112°C
98.5% min. (GC)
C18H32O2
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
02,IV,1754
15,556
Solubility in water: insoluble. Other solubilities: freely soluble in diethyl ether,miscible with dimethylformamide,soluble in ethanol,petroleum ether
CCCCCC=CCC=CCCCCCCCC(=O)O
280.45
CHEBI:17351
99%
60-33-3
Colorless to Yellow
229.0°C to 230.0°C (16.0 mmHg)
Authentic
Glass bottle
1.4687 to 1.4707
25 mL
0.9
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
OYHQOLUKZRVURQ-HZJYTTRNSA-N
(9Z,12Z)-octadeca-9,12-dienoic acid
5280450
280.45
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

EINECSNumber : 200-470-9

SDS
Documents

Documents

Promotions

Promotions