missing translation for 'onlineSavingsMsg'
Learn More

Linoleic acid, 95%

CAS: 60-33-3 | C18H32O2 | 280.452 g/mol

Supplier:  Thermo Scientific Chemicals L0794906

 View more versions of this product

Catalog No. AAL0794906


Only null left

Description

Description

Unsaturated μ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Unsaturated µ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.

Solubility
Miscible with dimethyl sulfoxide, ethanol, ether, petroleum ether, dimethylformamide, acetone and benzene. Insoluble in water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Linoleic acid
-12°C
230°C (16 mmHg)
Weak, Characteristic
1.4699
MFCD00064241
1727101
14,5505
Miscible with dimethyl sulfoxide,ethanol,ether,petroleum ether,dimethylformamide,acetone and benzene. Insoluble in water.
CCCCCC=CCC=CCCCCCCCC(=O)O
280.452
CHEBI:17351
95%
60-33-3
0.902
>110°C (230°F)
C18H32O2
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
5 g
Air sensitive
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
OYHQOLUKZRVURQ-HZJYTTRNSA-N
(9Z,12Z)-octadeca-9,12-dienoic acid
5280450
280.45

Chemical Identifiers

60-33-3
280.452
OYHQOLUKZRVURQ-HZJYTTRNSA-N
5280450
(9Z,12Z)-octadeca-9,12-dienoic acid
C18H32O2
MFCD00064241
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
CHEBI:17351
CCCCCC=CCC=CCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-470-9

RTECSNumber : RF9990000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only