missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linoleic acid, 70%

CAS: 60-33-3 | C18H32O2 | 280.45 g/mol

Supplier:  Thermo Scientific Chemicals 413190025

 View more versions of this product

Catalog No. AC413190025


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Linoleic acid
60-33-3
-5.0°C
0.9000g/mL
>112°C
2% max. >C18 (GC)
C18H32O2
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
2.5 kg
0.9
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
OYHQOLUKZRVURQ-HZJYTTRNSA-N
(9Z,12Z)-octadeca-9,12-dienoic acid
5280450
280.45
Technical
tech., 60%
197 to 203mg KOH/g
Yellow
229.0°C to 230.0°C (16.0 mmHg)
Authentic
Plastic drum
1.4687 to 1.4707
MFCD00064241
02, IV, 1754
15, 5560
Solubility in water: insoluble. Other solubilities: freely soluble in diethyl ether,miscible with dimethylformamide,soluble in ethanol,petroleum ether
CCCCCC=CCC=CCCCCCCCC(=O)O
280.45
CHEBI:17351
60%
Viscous Liquid

Chemical Identifiers

60-33-3
280.45
OYHQOLUKZRVURQ-HZJYTTRNSA-N
5280450
(9Z,12Z)-octadeca-9,12-dienoic acid
C18H32O2
MFCD00064241
linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
CHEBI:17351
CCCCCC=CCC=CCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-470-9

RTECSNumber : RF9990000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only