Beckman Coulter HarvestLine system liner kit comes with: one bottle rack and one funnel. This kit does not include the HarvestLine system liners. (Qty 1)
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Beckman Coulter HarvestLine system liner kit comes with: one bottle rack and one funnel. This kit does not include the HarvestLine system liners. (Qty 1)