missing translation for 'onlineSavingsMsg'
Learn More

ThermoHeat™ Liner with Heat Packs

Catalog No. 296355869
Change view
Click to view available options
Color:
Navy Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
296355869 Navy Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296355869 Supplier ThermoHeat™ Supplier No. 1070016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GL4 with heat packs channels and two heat packs

  • Long neck length navy blue outer shell with soft red lining to be worn under hard hats with channel heat packs
  • Three suspension tabs
  • Velcro chin closure

Specifications

Product Type Cold-weather Hard Hat Liner
Size Universal
Color Navy Blue
Disposable Reusable
Includes Hood with heat pack channels
For Use With (Application) Head protection from cold