Learn More
Thermo Scientific™ Lindberg/Blue M™ LGO Box Furnace, 0.6 cu. ft., vertical lift door, multi seg/multi prog/OTC
LGO Box Furnaces feature the latest technical advances in heating elements, insulation and temperature control, all integrated into a self-contained cabinet.
Supplier:  Thermo Scientific™ BF51732PBC1
Catalog No. 10549145
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Thermo Scientific™ LGO Box Furnaces feature the latest technical advances in heating elements, insulation and temperature control, all integrated into a self-contained cabinet. Furnaces feature proprietary LGO (light gauge overbend) heating elements and Moldatherm™ insulation for efficient and economical transfer of heat to chamber, with low exterior temperatures.
  • Variable heat-up rate eliminates thermal shock to materials with quick heat-up and cool-down rates
  • Choice of side hinge or vertical lift door
  • Air vent (1in. dia., top) and air inlet (0.375in. dia., rear) for inert atmosphere exchange (may experience some leakage at door)
  • Self-tuning, digital instrumentation for precise temperature setpoint and display
  • Platinel II™ thermocouple for long-term stability
  • 0.6 cu. ft. models feature vertical lift door; 2 cu. ft. models feature horizontal side swing door, hot side facing away from operator for protection
Microprocessor Control
  • Microprocessor-based self-tuning PID control (proportional, integral, derivative) provides optimum thermal process, prevents overshoot
  • Control panel designed for easy access and maintenance
  • Main power ON/OFF switch on control panel
  • Adjustable high-limit overtemperature protection
  • Simultaneous LED display of actual temperature vs. setpoint
  • Can be configured to display temperature in either °C or °F
  • Safety door switch interrupts power to heating element when door is opened; protects heating elements and minimizes exposure to end-user
  • Removable shelves for versatility
  • Moldatherm™ hearthplate supports load and prevents damage due to spillage
Digital, Single-Setpoint Controller
  • Single segment, single setpoint, one ramp to setpoint
Digital Single-Program, Multiple-Segment Programmable Controller
  • Single program with multiple segments for ramp (up and down) and dwell (timed hold) temperature control
Digital Multiple-Program, Multiple-Segment Programmable Controller
  • Available on models with
    P designation Multiple programs and segments for ramp (up and down) and dwell (timed hold) temperature control
Overtemperature Control (OTC)†
  • Adjustable digital overtemperature control, available on selected models with
    B suffix designation
  • Protects furnace and load in the event of primary control circuit failure
  • Overrides main controller and shuts off power to furnace if high limit is reached
  • Manual reset required for safety
  • Operates via magnetic contacts through signal from independent thermocouple
Flowmeter Option (FM) (Inert Atmosphere Only)
  • Available on selected models with
    FM designation
  • Gas flowmeter, adjustable, located on front control panel
  • Adjustable flowrate, range 1.0 to 10.0 cu. ft./hr. standard
  • Suitable for inert gas or airflow to chamber
  • Allows fresh air exchange for ashing applications Not suitable for combustible or volatile gases
Ordering Information:
Required power cord and hardwiring not included. Optional RS485 Digital Communications Port allows controller to be connected to a PC for remote monitoring and control of the furnace; up to 30 units can be connected to one PC.
†Specify Option
B for Overtemperature Control (OTC) when ordering.
Includes:
One two-part shelf (0.6 cu. ft. models have one shelf position at center position; 2.0 cu. ft. models have three shelf positions)
Specifications
Specifications
0.6 cu.ft.
100°C to 1200°C
4500 w
11 x 12 x 11 in. (27.9 x 33 x 17.8 cm)
23 in.
24 in.
33 cm
11 in.
17.8 cm
27.9 cm
208/240 V 50/60 Hz
50/60 Hz
16.4 L
Multisegment, Multiprogram, OTC
Digital Multi Seg/Multi Prog/OTC
23 x 24 x 27 in. (58.4 x 61 x 68.6 cm)
58.4
61 cm
27 in.
68.6 cm
11 in.
75 kg
208/240 V
Product Suggestions
Product Suggestions