missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers

Catalog No. 01257071
Change view
Click to view available options
Type:
Single-zone Controller, Digital
For Use With (Equipment):
Lindberg 1200C Tube Furnace
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
01257071 Single-zone Controller, Digital Lindberg 1200C Tube Furnace
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257071 Supplier Thermo Scientific™ Supplier No. CC58114A1*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Program segments into Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers for increased temperature accuracy.

Program segments into Thermo Scientific™ Lindberg/Blue M™ Box and Tube Furnace Controllers for increased temperature accuracy. Features overtemperature control and multiple segment configuration; designed to work with the 1200°C split-hinge and 1500°C box furnaces.

Features

  • Fully wired with advanced microprocessor-based digital control, solid-state power module, on/off circuit breaker and thermocouple input jacks for each zone
  • Built-in adjustable high-limit overtemperature protection
  • LED simultaneously displays actual temperature vs. setpoint in °C or °F
  • Designed for operation on 120, 208, or 240V 50/60Hz, single-phase line

Overtemperature Control

  • Adjustable digital control is factory installed
  • Protects furnace and load in the event of primary control circuit failure
  • Overrides main controller and shuts off power to furnace if high limit is reached
  • Must be manually reset for safety
  • Operates via magnetic contacts through a signal from an independent thermocouple

1200°C Three Zone, 16 Segment, One Programmable Controller for Split-hinge Furnaces (CC58434C)

  • Three microprocessor-based PID controls, single segment, single setpoint, one ramp to set point

1200°C Three Zone, 16 Segment, Three Programmable Controllers for Split-hinge Furnaces (CC58434PC)

  • Center Zone-One microprocessor-based PID programmable control, with one program of 16 segments for ramps (up and down) and eight dwells (timed holds)
  • End Zones-Two microprocessor-based PID control, single segment, single setpoint; microprocessors mimic the programmed profile of the center zone controller, but allow a digitally displayed offset up to 100°C

1200°C Digital, Single Setpoint Controllers for Split-hinge Furnaces (CC58114PBA, CC58114PA)

  • Includes one microprocessor-based PID control, single segment, single setpoint, one ramp to setpoint
  • CC58114PBA includes overtemperature control

1500°C Digital, Single Setpoint Controller for Box Furnaces (CC58125C)

  • Includes three microprocessor-based PID controls, single segment, single setpoint, one ramp to set point

1500°C Digital, Single Program, Multiple Segment Programmable Controllers for Box Furnaces (CC58125PC, CC58125PBC)

  • Advanced microprocessor-based PID control, single program with multiple segments for ramp and dwell temperature control
  • CC58125PBC includes overtemperature control

Specifications

For Use With (Equipment) Lindberg 1200C Tube Furnace
Temperature (Metric) 1200°C (Split-Hinge), 1500°C (Box Furnaces)
Type Single-zone Controller, Digital
Display Digital
Voltage 120 V
Amperage 30 A
Frequency 50/60 Hz