missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Linbro™ Liquisystem, Polypropylene Tubes
Used as Lab essentials
Supplier: MP Biomedicals™ 0961226C2
Description
- Storage System
- Sterile
- Store at room temperature
Specifications
| Each | |
| Polypropylene | |
| Sterile |
| Linbro Liquisystem | |
| 2°C to 8°C |