missing translation for 'onlineSavingsMsg'
Learn More

Linalool 96.0+%, TCI America™

Catalog No. L0048500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0048500ML 500 mL
L004825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0048500ML Supplier TCI America Supplier No. L0048500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-70-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00008906
InChI Key CDOSHBSSFJOMGT-UHFFFAOYSA-N
Synonym linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
PubChem CID 6549
ChEBI CHEBI:17580
IUPAC Name 3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C

Specifications

Color Yellow
Boiling Point 200°C
Quantity 500 mL
Formula Weight 154.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Linalool
EINECSNumber (2)-0249&(2)-0258
RTECSNumber RG5775000
TSCA Yes