missing translation for 'onlineSavingsMsg'
Learn More

Linalool Oxide (mixture of isomers) 97.0+%, TCI America™

Catalog No. l014325ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L014325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L014325ML Supplier TCI America Supplier No. L014325ML
Only null left

Chemical Identifiers

CAS 60047-17-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00053543
InChI Key BRHDDEIRQPDPMG-UHFFFAOYSA-N
Synonym Linalyl Oxide, 2-(2-Hydroxy-2-propyl)-5-methyl-5-vinyltetrahydrofuran, 2-(5-Methyl-5-vinyltetrahydro-2-furyl)-2-propanol
PubChem CID 22310
IUPAC Name 2-(5-ethenyl-5-methyloxolan-2-yl)propan-2-ol
SMILES CC1(CCC(O1)C(C)(C)O)C=C

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Boiling Point 78°C
Formula Weight 170.25
Percent Purity ≥97.0% (GC)
Chemical Name or Material Linalool Oxide (mixture of isomers)