missing translation for 'onlineSavingsMsg'
Learn More

Linalool, 97%

CAS: 78-70-6 | C10H18O | 154.253 g/mol

Supplier:  Thermo Scientific Chemicals A14424AP

 View more versions of this product

Catalog No. AAA14424AP


Only null left

Description

Description

Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.

Solubility
Soluble in water (1.589 g/L at 25°C).

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Linalool
0.865
76°C (168°F)
C10H18O
MFCD00008906
1721488
linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
CDOSHBSSFJOMGT-UHFFFAOYSA-N
3,7-dimethylocta-1,6-dien-3-ol
6549
154.25
Liquid
78-70-6
199°C to 200°C
97%
1.462
500 mL
14,5495
Soluble in water (1.589g/L at 25°C).
CC(=CCCC(C)(C=C)O)C
154.253
CHEBI:17580
97%

Chemical Identifiers

78-70-6
154.253
CDOSHBSSFJOMGT-UHFFFAOYSA-N
6549
3,7-dimethylocta-1,6-dien-3-ol
C10H18O
MFCD00008906
linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
CHEBI:17580
CC(=CCCC(C)(C=C)O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c

H227-H315-H317-H319

EINECSNumber : 201-134-4

RTECSNumber : RG5775000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only