missing translation for 'onlineSavingsMsg'
Learn More

Lime Water, Saturated, LabChem™

Catalog No. LC161602
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC161602 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC161602 Supplier LabChem Supplier No. LC161602
Only null left

Specifications

Color Clear, colorless
Format Liquid
Packaging Poly Bottle
Quantity 1 L
Storage Requirements Room Temperature
Type Lime Water