missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lignoceric Acid 96.0+%, TCI America™
$180.81 - $1457.70
Chemical Identifiers
| CAS | 557-59-5 |
|---|---|
| Molecular Formula | C24H48O2 |
| Molecular Weight (g/mol) | 368.646 |
| MDL Number | MFCD00002810 |
| InChI Key | QZZGJDVWLFXDLK-UHFFFAOYSA-N |
| Synonym | lignoceric acid, unii-rk3vcw5y1l, n-tetracosanoic acid, rk3vcw5y1l, n-tetracosanoate, lignozerinsaeure, tetracosansaeure, carnaubic acid, tetracosoic acid, tetracosanic acid |
| PubChem CID | 11197 |
| ChEBI | CHEBI:28866 |
| IUPAC Name | tetracosanoic acid |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCC(=O)O |
Chemical Identifiers
| 557-59-5 | |
| 368.646 | |
| QZZGJDVWLFXDLK-UHFFFAOYSA-N | |
| 11197 | |
| tetracosanoic acid |
| C24H48O2 | |
| MFCD00002810 | |
| lignoceric acid, unii-rk3vcw5y1l, n-tetracosanoic acid, rk3vcw5y1l, n-tetracosanoate, lignozerinsaeure, tetracosansaeure, carnaubic acid, tetracosoic acid, tetracosanic acid | |
| CHEBI:28866 | |
| CCCCCCCCCCCCCCCCCCCCCCCC(=O)O |
Specifications
| 557-59-5 | |
| White-Yellow | |
| MFCD00002810 | |
| lignoceric acid, unii-rk3vcw5y1l, n-tetracosanoic acid, rk3vcw5y1l, n-tetracosanoate, lignozerinsaeure, tetracosansaeure, carnaubic acid, tetracosoic acid, tetracosanic acid | |
| CCCCCCCCCCCCCCCCCCCCCCCC(=O)O | |
| 368.646 | |
| CHEBI:28866 | |
| ≥96.0% (T) | |
| Lignoceric Acid |
| 84°C | |
| C24H48O2 | |
| 1 g | |
| QZZGJDVWLFXDLK-UHFFFAOYSA-N | |
| tetracosanoic acid | |
| 11197 | |
| 368.65 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : Yes