missing translation for 'onlineSavingsMsg'
Learn More

Lignoceric Acid 96.0+%, TCI America™

Catalog No. T00761G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00761G 1 g
T00765G 5 g
T007625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T00761G Supplier TCI America Supplier No. T00761G

Chemical Identifiers

CAS 557-59-5
Molecular Formula C24H48O2
Molecular Weight (g/mol) 368.646
MDL Number MFCD00002810
InChI Key QZZGJDVWLFXDLK-UHFFFAOYSA-N
Synonym lignoceric acid, unii-rk3vcw5y1l, n-tetracosanoic acid, rk3vcw5y1l, n-tetracosanoate, lignozerinsaeure, tetracosansaeure, carnaubic acid, tetracosoic acid, tetracosanic acid
PubChem CID 11197
ChEBI CHEBI:28866
IUPAC Name tetracosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 1 g
Formula Weight 368.65
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lignoceric Acid
EINECSNumber (2)-0608
TSCA Yes