missing translation for 'onlineSavingsMsg'
Learn More

Lignin (Dealkaline), TCI America™

Catalog No. p-7131790
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L004525G 25 g
L0045500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L004525G Supplier TCI America Supplier No. L004525G
Only null left

Chemical Identifiers

CAS 9005-53-2
MDL Number MFCD00147441

Specifications

Color Black-Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Lignin (Dealkaline)
EINECSNumber (8)-0215
TSCA Yes