Learn More
BINDER™ Light Module for Plants
Description
LED plant light module with 16 LED strip lights
Includes:
- 3-channel controller
- Fresh air pump
- Independent power supply
Specifications
Specifications
| For Use With (Equipment) | KB PRO 260, KB PRO 470, KB PRO 720, KB PRO 1060, KB PRO 1600, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 |
| Material | Glass |
| Type | Lighting |
| Features | 16 LED Strip Lights |