Learn More
Light Mineral Oil, NF, Spectrum™ Chemical
M1293, 8042-47-5
Supplier:  Spectrum Chemical Mfg Cor M12934LTGLCS4
 View more versions of this product
Catalog No. 18603892
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Light Mineral Oil, NF is used as a vehicle and humectant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
8042-47-5
100%
Amber Glass Bottle
4 x 4 L
Chemical Identifiers
8042-47-5
Product Suggestions
Product Suggestions