missing translation for 'onlineSavingsMsg'
Learn More

Light Green SF Yellowish, Spectrum™ Chemical

Catalog No. 18611289
Change view
Click to view available options
Quantity:
10 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611289 5 kg Amber Glass Bottle
18611286 10 g Amber Glass Bottle
18611287 100 g Amber Glass Bottle
18611285 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611289 Supplier Spectrum Chemical Mfg Cor Supplier No. LI1055KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Light Green SF Yellowish is a triarylmethane dye primarily used in histology for staining collagen.

  • Chemical Name: Light Green SF Yellowish
  • Formula: C37H34N2O9S3Na2
  • Molecular Weight: 792.86 g/mole
  • Physical State: Powder
  • Solubility: Soluble in water
  • Applications: Used in histology for staining collagen; also used for dyeing wool
  • CAS Number: 5141-20-8
  • CI Number: 42095

Chemical Identifiers

CAS 5141-20-8

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Dark Violet
Quantity 5 kg
Molecular Weight (g/mol) 792.86
Grade Reagent
Chemical Name or Material Light Green SF Yellowish

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.