missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Light Green SF, Yellowish, pure, high purity biological stain

Catalog No. AC229770100
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229770100 10 g Glass bottle
AC229771000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229770100 Supplier Thermo Scientific Chemicals Supplier No. 229770100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5141-20-8 | C37H34N2Na2O9S3 | 792.844 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5141-20-8
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012121
InChI Key DGOBMKYRQHEFGQ-UHFFFAOYSA-L
Synonym Acid Green 5, C.I. 42095
PubChem CID 21223
ChEBI CHEBI:87065
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 288.0°C
Absorption Ratio 0.96 to 1.16
Beilstein 14, 856
Merck Index 15, 5538
Solubility Information Solubility in water: 200g/L water (20°C). Other solubilities: soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 629 to 634nm (H2O, 5mg/L, 1cm)
Formula Weight 792.84
Loss on Drying 10% max. (1 g, 110°C, 2 hrs)
Percent Purity ≥60% (TiCl3-titr.) (on substance as is)
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Purple
Quantity 10 g
Physical Form Powder, Crystals or Crystalline Powder
Chemical Name or Material Light Green SF, Yellowish, Pure
Show More Show Less

RUO – Research Use Only