missing translation for 'onlineSavingsMsg'
Learn More

Light Green SF Yellowish (formerly Food Green No.2), TCI America™

Catalog No. L01411G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L01411G Supplier TCI America Supplier No. L01411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[as an indicator for assay of Food Yellow No.4 (Tartrazine)]

Chemical Identifiers

CAS 5141-20-8
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012121
InChI Key DGOBMKYRQHEFGQ-UHFFFAOYSA-L
PubChem CID 21223
ChEBI CHEBI:87065
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Purple/Red
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 792.84
Grade Indicator
Chemical Name or Material Light Green SF Yellowish (formerly Food Green No.2) [as an indicator for assay of Food Yellow No.4 (
EINECSNumber (5)-4374
RTECSNumber BQ4900000
TSCA Yes