missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Light Green SF, Yellowish
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Bio. Stain Comm. Certification | Pass Test |
| Dye Content | REPORT% |
| Vapor Pressure | Vapor Pressure: Negligible |
| Decomposition Information | 288°C |
| Beilstein | 14, 856 |
| Merck Index | 15, 5538 |
| Quantity | 25 g |
| Formula Weight | 792.84 |
| Grade | Extra Pure |
| Chemical Name or Material | Light Green SF, Yellowish |
RUO – Research Use Only