missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Light Green, 0.2% (w/v), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Light Green |
| CAS | 5141-20-8, 7732-18-5 |
| CAS Min % | 98.0, 0.2 |
| CAS Max % | 100.0, 0.2 |
| Concentration or Composition (by Analyte or Components) | 0.2% (w/v) |
| Packaging | Natural Poly Bottle |
| Quantity | 500 mL |