Learn More
Celltreat Scientific Products Light Enhancing Sealing Film, White, Non-sterile
Catalog No. 5000144347
Encompass
Black and white film is suitable for application on the top or bottom of black and white multiple well platesEasily cover clear bottom black or white plates to prepare for top-reading analytical equipmentBlack film absorbs stray light and fluorescence, dampening background noise and improving signal clarityWhite film reflects weak light and fluorescent signals to enhance output readingsPlace black film on top of black plates to fully shield light-sensitive samples
Catalog No. 5000144347 Supplier Celltreat Scientific Products Supplier No. 229140
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Black and white film is suitable for application on the top or bottom of black and white multiple well platesEasily cover clear bottom black or white plates to prepare for top-reading analytical equipmentBlack film absorbs stray light and fluorescence, dampening background noise and improving signal clarityWhite film reflects weak light and fluorescent signals to enhance output readingsPlace black film on top of black plates to fully shield light-sensitive samples