Learn More
International Enviroguard™ Light Blue SMS Lab Coat with 3 Pockets, Knit Wrists
SMS lab coat protects against dust, dirt, grime, and dry particulates in nonhazardous settings.
Supplier: International Enviroguard™ K2248BM
Description
Light blue SMS lab coat protects against dust, dirt, grime, and other dry particulates in non-hazardous environments. Fabric also helps protect against light liquid splashes and sprays. Fabric is breathable to help keep you cool, so you can work comfortably over long hours. With three roomy pockets, you can store eyeglasses, pens, and other small instruments to keep with you on the go.
- Three-layer SMS is lightweight and breathable to help prevent heat stress
- Knit wrists and knit collar
- 1 left chest pocket and 2 hip pockets
- Possible applications include pharmaceuticals, labs, healthcare, controlled environments, food processing, and more
Specifications
| General Purpose Lab Coat | |
| Unisex | |
| Blue | |
| Knit | |
| Non-sterile | |
| 3 | |
| Knit | |
| K2248B-M | |
| Not Autoclavable |
| Medium | |
| SMS | |
| Snaps | |
| Single-use | |
| Below Hip-length | |
| Blue SMS Lab Coat with 3 pockets, knit wrists, and knit collar | |
| Yes | |
| Serged | |
| 30/Cs. |