Learn More
Tradewinds Direct Inc BioX 180mm Cell Lifter. Case of 100
Catalog No. 502274282
Encompass
BioX disposable polyethylene sterile cell lifter (handle length: 180mm, blade length: 20mm). Lifters come individually wrapped. 100/case, price/case
Catalog No. 502274282 Supplier Tradewinds Direct Inc Supplier No. BX702180
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BioX disposable polyethylene sterile cell lifter (handle length: 180mm, blade length: 20mm). Lifters come individually wrapped. 100/case, price/case