Learn More
LifeSign Easicult™ Combi DFS Item
Dip-slide method with two media for the culture of yeast and fungi and most aerobic bacteria
Supplier:  LifeSign 4060
Catalog No. 23-264781
Only null left
Explore available promotions
Description
Description
Dip-slide method with two media for the culture of yeast and fungi and most aerobic bacteria
  • Prolongs the lifetime of industrial fluids by allowing adjustment of the microbial level
  • Prevents microbial matter from clogging filters and pumps
  • Prevents machine, tank, and pipe corrosion by minimizing the amount of microbial metabolites and by stabilizing the amount of lubrication
  • Optimizes the level of biocides by monitoring the microbial counts
  • Prevents process stoppages caused by microbes
  • Improves the hygienic conditions of working areas
Specifications
Specifications
Solid
For monitoring microbes in industrial fluids
10 Tests
United States
Slide
Product Suggestions
Product Suggestions